siRNA Details
| Gene Symbol: Gad1os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040496.1 | 1 | 90 | |||
| GenBank mRNA | AK005124.1 | - | 1 | 91 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY705319.1 | - | 1 | 88 | Chr. 2 - 70489958 - 70563355 [-] on Build GRCm38 | |

Gad1os
Genomic Map