siRNA Details
| Gene Symbol: Rbm46os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040381.1 | 4 | 2328 | |||
| NR_040382.1 | 4 | 2319 | ||||
| GenBank mRNA | AK015378.1 | - | 543 | |||
| AK161442.1 | - | 4 | 2329 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CA463122.1 | - | 4 | 468 | Chr. 3 - 82876705 - 82942955 [+] on Build GRCm38 | |

Rbm46os
Genomic Map