siRNA Details
| Gene Symbol: Rbm46os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040381.1 | 4 | 2774 | |||
| NR_040382.1 | 4 | 2765 | ||||
| GenBank mRNA | AK015378.1 | - | 989 | |||
| AK161442.1 | - | 4 | 2775 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AV271192.2 | - | 1 | 156 | Chr. 3 - 82942996 - 82943635 [+] on Build GRCm38 | |

Rbm46os
Genomic Map