siRNA Details
| Gene Symbol: Rbm46os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015378.1 | - | 504 | |||
| CJ047138.1 | - | 427 | ||||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CA463122.1 | - | 3, 4 | 429 | Chr. 3 - 82876705 - 82942955 [+] on Build GRCm38 | |

Rbm46os
Genomic Map