siRNA Details
| Gene Symbol: 4930423M02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038183.1 | 4 | 828 | |||
| GenBank mRNA | AK015191.1 | - | 4 | 831 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DV660962.1 | - | 3 | 691 | Chr. 4 - 5635835 - 5641896 [+] on Build GRCm38 | |

4930423M02Rik
Genomic Map