siRNA Details
| Gene Symbol: 4930423M02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038183.1 | 4 | 528 | |||
| GenBank mRNA | AK015191.1 | - | 4 | 531 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930423M02Rik| Gene Symbol: 4930423M02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038183.1 | 4 | 528 | |||
| GenBank mRNA | AK015191.1 | - | 4 | 531 | ||