siRNA Details
| Gene Symbol: 4933429O19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033783.1 | 3 | 377 | |||
| GenBank mRNA | AK015198.1 | - | 3 | 333 | ||
| AK016985.1 | - | 3 | 367 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY716669.1 | - | 3 | 367 | Chr. 14 - 48878884 - 48886191 [+] on Build GRCm38 | |

4933429O19Rik
Genomic Map