siRNA Details
| Gene Symbol: Stag1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK034136.1 | - | 3317 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB079646.2 | - | 1 | 374 | Chr. 9 - 100958710 - 100959372 [+] on Build GRCm38 | |

Stag1
Genomic Map