siRNA Details
| Gene Symbol: Gm38416 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK028839.1 | - | 101 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB613169.1 | - | 1 | 101 | Chr. 8 - 117681622 - 117683972 [+] on Build GRCm38 | |

Gm38416
Genomic Map