siRNA Details
| Gene Symbol: Gm9962 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033504.1 | 3 | 563 | |||
| GenBank mRNA | AK049950.1 | - | 3 | 564 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm9962| Gene Symbol: Gm9962 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033504.1 | 3 | 563 | |||
| GenBank mRNA | AK049950.1 | - | 3 | 564 | ||