siRNA Details
| Gene Symbol: 4930539C22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015994.1 | - | 9 | 954 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930539C22Rik| Gene Symbol: 4930539C22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015994.1 | - | 9 | 954 | ||