siRNA Details
| Gene Symbol: 4930579D09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016315.1 | - | 2 | 415 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715954.1 | - | 2 | 418 | Chr. 6 - 143533963 - 143535510 [-] on Build GRCm38 | |

4930579D09Rik
Genomic Map