siRNA Details
| Gene Symbol: 4933433G19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045851.1 | 7 | 643 | |||
| GenBank mRNA | AK017041.1 | - | 7 | 646 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4933433G19Rik| Gene Symbol: 4933433G19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045851.1 | 7 | 643 | |||
| GenBank mRNA | AK017041.1 | - | 7 | 646 | ||