siRNA Details
| Gene Symbol: 4933433G19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045851.1 | 7 | 667 | |||
| GenBank mRNA | AK017041.1 | - | 7 | 670 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY716725.1 | - | 668 | Chr. - - [-] on Build | ||

4933433G19Rik
Genomic Map