siRNA Details
| Gene Symbol: 4933433H22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045458.1 | 4 | 745 | |||
| GenBank mRNA | AK017043.1 | - | 4 | 743 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4933433H22Rik| Gene Symbol: 4933433H22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045458.1 | 4 | 745 | |||
| GenBank mRNA | AK017043.1 | - | 4 | 743 | ||