siRNA Details
| Gene Symbol: 1810053B23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040486.1 | 4 | 1028 | |||
| NR_040488.1 | 4 | 870 | ||||
| NR_040489.1 | 4 | 868 | ||||
| GenBank mRNA | AK007854.1 | - | 4 | 1029 | ||
| AK160158.1 | - | 4 | 873 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1810053B23Rik| Gene Symbol: 1810053B23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040486.1 | 4 | 1028 | |||
| NR_040488.1 | 4 | 870 | ||||
| NR_040489.1 | 4 | 868 | ||||
| GenBank mRNA | AK007854.1 | - | 4 | 1029 | ||
| AK160158.1 | - | 4 | 873 | |||