siRNA Details
| Gene Symbol: Gm16677 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK046150.1 | - | 611 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm16677| Gene Symbol: Gm16677 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK046150.1 | - | 611 | |||