siRNA Details
| Gene Symbol: Gm20756 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040771.1 | 2 | 245 | |||
| GenBank mRNA | AK165854.1 | - | 2 | 246 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY733948.1 | - | 2 | 246 | Chr. 6 - 25931426 - 25943142 [+] on Build GRCm38 | |

Gm20756
Genomic Map