siRNA Details
| Gene Symbol: Magef1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK018285.1 | - | 1 | 967 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB469018.2 | - | 1 | 180 | Chr. 16 - 21331907 - 21332575 [-] on Build GRCm38 | |

Magef1
Genomic Map