siRNA Details
| Gene Symbol: Gm4872 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_073371.1 | 3 | 1285 | |||
| GenBank mRNA | AK052410.1 | - | 3 | 1286 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm4872| Gene Symbol: Gm4872 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_073371.1 | 3 | 1285 | |||
| GenBank mRNA | AK052410.1 | - | 3 | 1286 | ||