siRNA Details
Gene Symbol: Mfap3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK017269.1 | - | 1 | 640 | ||
AK122332.1 | - | 2 | 4597 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mfap3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK017269.1 | - | 1 | 640 | ||
AK122332.1 | - | 2 | 4597 |