siRNA Details
| Gene Symbol: 9430032L10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020454.1 | - | 1 | 152 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY721705.1 | - | 1 | 152 | Chr. 17 - 37081225 - 37081884 [+] on Build GRCm38 | |

9430032L10Rik
Genomic Map