siRNA Details
| Gene Symbol: 9430031J08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020451.1 | - | 1 | 269 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY721697.1 | - | 1 | 269 | Chr. 15 - 33557052 - 33557665 [+] on Build GRCm38 | |

9430031J08Rik
Genomic Map