siRNA Details
| Gene Symbol: Dtymk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK080078.1 | - | 1, 2 | 27 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB637195.1 | - | 1, 2 | 23 | Chr. 1 - 93798440 - 93799349 [+] on Build GRCm38 | |

Dtymk
Genomic Map