siRNA Details
| Gene Symbol: Gm16619 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK079684.1 | - | 3 | 534 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB634338.1 | - | 3 | 534 | Chr. 16 - 49800533 - 49804292 [+] on Build GRCm38 | |

Gm16619
Genomic Map