siRNA Details
Gene Symbol: 6430710C18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_102348.1 | 2 | 209 | |||
NR_102349.1 | 2 | 209 | ||||
GenBank mRNA | AK018302.1 | - | 2 | 165 | ||
AK019889.1 | - | 2 | 209 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 6430710C18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_102348.1 | 2 | 209 | |||
NR_102349.1 | 2 | 209 | ||||
GenBank mRNA | AK018302.1 | - | 2 | 165 | ||
AK019889.1 | - | 2 | 209 |