siRNA Details
| Gene Symbol: 6430710C18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102348.1 | 3 | 469 | |||
| NR_102349.1 | 4 | 568 | ||||
| GenBank mRNA | AK018302.1 | - | 3 | 425 | ||
| AK019889.1 | - | 4 | 568 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

6430710C18Rik| Gene Symbol: 6430710C18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102348.1 | 3 | 469 | |||
| NR_102349.1 | 4 | 568 | ||||
| GenBank mRNA | AK018302.1 | - | 3 | 425 | ||
| AK019889.1 | - | 4 | 568 | |||