siRNA Details
| Gene Symbol: Galnt14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK144036.1 | - | 1 | 3013 | ||
| AK165509.1 | - | 1 | 3013 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB747279.1 | - | 1 | 353 | Chr. 17 - 73514715 - 73515095 [-] on Build GRCm38 | |

Galnt14
Genomic Map