siRNA Details
| Gene Symbol: AI256396 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK046686.1 | - | 2507 | |||
| AK134520.1 | - | 1442 | ||||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BX512726.1 | - | 1 | 405 | Chr. 6 - 100809553 - 100810075 [+] on Build GRCm38 | |

AI256396
Genomic Map