siRNA Details
| Gene Symbol: AI256396 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK046686.1 | - | 2734 | |||
| AK134520.1 | - | 1669 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

AI256396| Gene Symbol: AI256396 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK046686.1 | - | 2734 | |||
| AK134520.1 | - | 1669 | ||||