siRNA Details
| Gene Symbol: C030009O12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK021058.1 | - | 1 | 45 | ||
| AK045829.1 | - | 1 | 469 | |||
| AK140124.1 | - | 1 | 519 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C030009O12Rik| Gene Symbol: C030009O12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK021058.1 | - | 1 | 45 | ||
| AK045829.1 | - | 1 | 469 | |||
| AK140124.1 | - | 1 | 519 | |||