siRNA Details
| Gene Symbol: C030007I01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK021055.1 | - | 1 | 270 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY728118.1 | - | 1 | 270 | Chr. 5 - 130844507 - 130844900 [+] on Build GRCm38 | |

C030007I01Rik
Genomic Map