siRNA Details
| Gene Symbol: A930028O11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020901.1 | - | 1 | 929 | ||
| BB280937.1 | - | 1 | 119 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A930028O11Rik| Gene Symbol: A930028O11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020901.1 | - | 1 | 929 | ||
| BB280937.1 | - | 1 | 119 | |||