siRNA Details
| Gene Symbol: Gm33867 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK134227.1 | - | 11 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB619687.1 | - | 11 | Chr. 3 - 143743377 - 143825205 [+] on Build GRCm38 | ||

Gm33867
Genomic Map