siRNA Details
| Gene Symbol: 1700061D13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006849.1 | - | 1 | 315 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY706830.1 | - | 1 | 312 | Chr. 5 - 105866373 - 105866775 [-] on Build GRCm38 | |

1700061D13Rik
Genomic Map