siRNA Details
| Gene Symbol: 1700061A03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006847.1 | - | 1 | 33 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY706828.1 | - | 1 | 32 | Chr. 19 - 5488744 - 5489202 [-] on Build GRCm38 | |

1700061A03Rik
Genomic Map