siRNA Details
| Gene Symbol: Gm32478 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC042796.1 | - | 2 | 968 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm32478| Gene Symbol: Gm32478 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC042796.1 | - | 2 | 968 | ||