siRNA Details
| Gene Symbol: 1600015H23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK005466.1 | - | 1 | 304 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY705668.1 | - | 1 | 304 | Chr. 11 - 109344286 - 109344633 [+] on Build GRCm38 | |

1600015H23Rik
Genomic Map