siRNA Details
| Gene Symbol: Gm16712 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_108021.1 | 1 | 112 | |||
| GenBank mRNA | AK146869.1 | - | 1 | 93 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY740036.1 | - | 1 | 93 | Chr. 17 - 55956033 - 55959381 [-] on Build GRCm38 | |

Gm16712
Genomic Map