siRNA Details
| Gene Symbol: Gm12159 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045100.1 | 3 | 992 | |||
| GenBank mRNA | AK041275.1 | - | 3 | 993 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB226319.2 | - | 1 | 253 | Chr. 11 - 45102872 - 45103574 [-] on Build GRCm38 | |

Gm12159
Genomic Map