siRNA Details
| Gene Symbol: Gm13415 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK037056.1 | - | 3 | 344 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB630131.1 | - | 3 | 341 | Chr. 2 - 24442836 - 24462948 [+] on Build GRCm38 | |

Gm13415
Genomic Map