siRNA Details
| Gene Symbol: Itprip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK014762.1 | - | 1 | 1118 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CK626294.1 | - | 1 | 214 | Chr. 19 - 47917793 - 47918298 [-] on Build GRCm38 | |

Itprip
Genomic Map