siRNA Details
| Gene Symbol: 4921504P20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK014816.1 | - | 1 | 655 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY714338.1 | - | 1 | 655 | Chr. 4 - 129460551 - 129461524 [-] on Build GRCm38 | |

4921504P20Rik
Genomic Map