siRNA Details
| Gene Symbol: C530044C16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045984.1 | 4 | 384 | |||
| GenBank mRNA | AK043004.1 | - | 4 | 385 | ||
| AK049716.1 | - | 2 | 199 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB640265.1 | - | 4 | 385 | Chr. 6 - 50776115 - 50814334 [+] on Build GRCm38 | |

C530044C16Rik
Genomic Map