siRNA Details
| Gene Symbol: C530044C16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045984.1 | 5 | 767 | |||
| GenBank mRNA | AK043004.1 | - | 5 | 768 | ||
| AK049716.1 | - | 3 | 1014 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB251594.2 | - | 1 | 84 | Chr. 6 - 50814339 - 50814893 [+] on Build GRCm38 | |

C530044C16Rik
Genomic Map