siRNA Details
| Gene Symbol: C530044C16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045984.1 | 5 | 1149 | |||
| GenBank mRNA | AK043004.1 | - | 5 | 1150 | ||
| AK049716.1 | - | 3 | 1396 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C530044C16Rik| Gene Symbol: C530044C16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045984.1 | 5 | 1149 | |||
| GenBank mRNA | AK043004.1 | - | 5 | 1150 | ||
| AK049716.1 | - | 3 | 1396 | |||