siRNA Details
| Gene Symbol: Lyrm7os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040284.1 | 2 | 353 | |||
| GenBank mRNA | AK087691.1 | - | 2 | 354 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lyrm7os| Gene Symbol: Lyrm7os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040284.1 | 2 | 353 | |||
| GenBank mRNA | AK087691.1 | - | 2 | 354 | ||