siRNA Details
| Gene Symbol: Lyrm7os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040284.1 | 3 | 685 | |||
| GenBank mRNA | AK087691.1 | - | 3 | 686 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB553409.2 | - | 2 | 440 | Chr. 11 - 54821328 - 54824967 [+] on Build GRCm38 | |

Lyrm7os
Genomic Map