siRNA Details
Gene Symbol: Zfp229 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK147958.1 | - | 624 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB759550.1 | - | 6 | 491 | Chr. 17 - 21733863 - 21769342 [+] on Build GRCm38 |
