siRNA Details
| Gene Symbol: 4930505M18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015707.1 | - | 2 | 251 | ||
| AK019701.1 | - | 2 | 224 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715465.1 | - | 2 | 221 | Chr. 7 - 37669827 - 37671549 [+] on Build GRCm38 | |

4930505M18Rik
Genomic Map